Learn More
Cytek Biosciences Inc Cell Stimulation Cocktail (500X) | 400 µL (4 x 100 µL)
Catalog No. NC1807763
Encompass_Preferred
Cell Stimulation Cocktail (500X) is a ready to use cocktail that can be used for polyclonal cell activation. It is composed of the phorbol ester, PMA (Phorbol 12-Myristate 13-Acetate), a calcium ionophore (Ionomycin), and the protein transport inhibitors Brefeldin A and Monensin. This Cell Stimulation Cocktail (500X) can be used to elicit cytokine production and retention in a variety of cell types allowing for detection of intracellular protein by flow cytometry.
Catalog No. NC1807763 Supplier Cytek Biosciences Inc Supplier No. TNB4975UL400
Add to Cart
Add to Cart
Cell Stimulation Cocktail (500X) is a ready to use cocktail that can be used for polyclonal cell activation. It is composed of the phorbol ester, PMA (Phorbol 12-Myristate 13-Acetate), a calcium ionophore (Ionomycin), and the protein transport inhibitors Brefeldin A and Monensin. This Cell Stimulation Cocktail (500X) can be used to elicit cytokine production and retention in a variety of cell types allowing for detection of intracellular protein by flow cytometry.