Learn More
Chemglass Life Sciences Cell Strainer, 100µm, Individually Wrapped, Color Coded Yellow, Sterile
SDP
Catalog No. 501897579
Encompass
¿ Sample filtration ¿ Sample preparation for flow cytometer ¿ Isolate primary cells to consistently obtain a uniform single-cell tissue suspension ¿ Non-pyrogenic, DNase- / RNase-free, non-cytotoxicityCell strainer can be used to obtain uniform single cell suspension which is compatible with 50mL centrifuge tubes of most popular brands on the market. Frames are made from polypropylene with nylon strainers. Individually packaged.
Catalog No. 50-189-7579 Supplier Chemglass Life Sciences Supplier No. CGE2007100
¿ Sample filtration ¿ Sample preparation for flow cytometer ¿ Isolate primary cells to consistently obtain a uniform single-cell tissue suspension ¿ Non-pyrogenic, DNase- / RNase-free, non-cytotoxicityCell strainer can be used to obtain uniform single cell suspension which is compatible with 50mL centrifuge tubes of most popular brands on the market. Frames are made from polypropylene with nylon strainers. Individually packaged.