Learn More
Chemglass Life Sciences Cell Strainer, 100µm, Individually Wrapped, Color Coded Yellow, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN2007100
Encompass_Preferred
Cell strainer can be used to obtain uniform single cell suspension which is compatible with 50mL centrifuge tubes of most popular brands on the market. Frames are made from polypropylene with nylon strainers. Individually packaged. Sample filtration. Sample preparation for flow cytometer. Isolate primary cells to consistently obtain a uniform single-cell tissue suspension. Non-pyrogenic, DNase- / RNase-free, non-cytotoxicity.
Catalog No. 50-189-7818