Learn More
Research Products International Corp Cell Strainers, 100um Mesh Size, Tab Style, Yellow, 50 per Pack
SDP
Catalog No. 501367929
Encompass
These Cell Strainers are designed to fit standard 50ml conical tubes. Ideal for isolating primary cells to obtain single cell suspensions. Also can be used in sample preparation filter debris from solution prior to Cytometry procedures. Available in three different mesh sizes to accommodate various applications. Cell Strainers are supplied, sterile, non-pyrogenic and individually wrapped. Cell Strainers are color coded for easy identification.
Catalog No. 50-136-7929 Supplier Research Products International Corp Supplier No. 162436
These Cell Strainers are designed to fit standard 50ml conical tubes. Ideal for isolating primary cells to obtain single cell suspensions. Also can be used in sample preparation filter debris from solution prior to Cytometry procedures. Available in three different mesh sizes to accommodate various applications. Cell Strainers are supplied, sterile, non-pyrogenic and individually wrapped. Cell Strainers are color coded for easy identification.