Learn More
MTC Bio 20µm cell strainer, red sterile, individually wrapped, w/ 1 reducing adapter, 50/pk
Supplier:  MTC Bio C7020
Encompass_Preferred
The SureStrain™ Premium Cell Strainer system is a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in eight color-coded porosities: 5µm (Orange), 10µm (Teal), 20µm (Red), 30µm (Purple), 40μm (blue), 50µm (grey) 70μm (white), and 100μm (yellow). The advanced frame/housing design also contributes to superior performance and reduced processing time, by eliminating the “air-lock” effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers.• Advanced design – faster flow through• Compatible with 6-well plates• Interlocking and stackable• Sterile; RNase, DNase, pyrogen and cytotoxin free• Four 1.5mm feet for straining into flat plates and dishes• Designed for use with 50mL tubes Adapter for 15mL tubes included**Patent Pending
Catalog No. NC3737143