Learn More
Bellco Glass, Inc. Cell Strainer, 40um, Blue
SDP
Catalog No. 502330188
Encompass_Preferred
DescriptionCell Strainers can be used for many applications including isolating cells and removing clumps or debrisPolypropylene housing with nylon mesh filter eliminated the need to fabricate filters from gauze, paper, nylon mesh, or other materialsExtended lip on housing enables aseptic handling with forcepsElongated gripping tab on housing for easy handlingCell strainers are designed to universally fit most brands of 50mL conical tubesUnique bulk packaging option available Pack Description10/BagCase Quantity50SterileYesNon-pyrogenicYesPrimary ColorBluePrimary MaterialPolypropyleneGraduatedNoRNase/DNase FreeYes
Catalog No. 50-233-0188 Supplier Bellco Glass, Inc. Supplier No. 229482000
DescriptionCell Strainers can be used for many applications including isolating cells and removing clumps or debrisPolypropylene housing with nylon mesh filter eliminated the need to fabricate filters from gauze, paper, nylon mesh, or other materialsExtended lip on housing enables aseptic handling with forcepsElongated gripping tab on housing for easy handlingCell strainers are designed to universally fit most brands of 50mL conical tubesUnique bulk packaging option available Pack Description10/BagCase Quantity50SterileYesNon-pyrogenicYesPrimary ColorBluePrimary MaterialPolypropyleneGraduatedNoRNase/DNase FreeYes