Learn More
Andwin Scientific CELL STRAINER 40UM 10/CS
SDP
Catalog No. 50727399
Encompass_Preferred
The 40 m Falcon Cell Strainers feature nylon mesh that offers users more even cell suspensions. This product can also be utilized for dissociating clumped cells in separation for cell counting or sub-culture, separating enzymatically dissociated cells from undigested tissue for the initiation of primary cell growing, tissue processing such as fixation, dehydration, and staining.The strainer is suitable to work with 50mL conical tubes. This is sold in a case of 10.
Catalog No. 50-727-399 Supplier Andwin Scientific Supplier No. 6475110
The 40 m Falcon Cell Strainers feature nylon mesh that offers users more even cell suspensions. This product can also be utilized for dissociating clumped cells in separation for cell counting or sub-culture, separating enzymatically dissociated cells from undigested tissue for the initiation of primary cell growing, tissue processing such as fixation, dehydration, and staining.The strainer is suitable to work with 50mL conical tubes. This is sold in a case of 10.