Learn More
Research Products International Corp Cell Strainer, 40µm Mesh, Purple, 100 per Package
SDP
Catalog No. 501367949
Encompass
These Cell Strainers are designed to fit standard 50ml conical tubes. Ideal for isolating primary cells to obtain single cell suspensions. Also can be used in sample preparation filter debris from solution prior to Cytometry procedures.
Strong nylon mesh for optimal performance in a variety of applications. Evenly spaced mesh pores providing consistent and reliable result. Polypropylene frame features a molded tab for easy handling.
Cell Strainers are supplied sterile, non-pyrogenic and individually wrapped.
Catalog No. 50-136-7949 Supplier Research Products International Corp Supplier No. 162425100
These Cell Strainers are designed to fit standard 50ml conical tubes. Ideal for isolating primary cells to obtain single cell suspensions. Also can be used in sample preparation filter debris from solution prior to Cytometry procedures.
Strong nylon mesh for optimal performance in a variety of applications. Evenly spaced mesh pores providing consistent and reliable result. Polypropylene frame features a molded tab for easy handling.
Cell Strainers are supplied sterile, non-pyrogenic and individually wrapped.