Learn More
Cole-Parmer Argos Technologies Cell Strainer, 40 um, Blue, Sterile, Individual Wrap, 50/Cs
Catalog No. 501960595 Shop All Cole-Parmer Products
Encompass_Preferred
Pair these cell strainers directly with your 50-mL centrifuge tubes. Choose from 40, 70 or 100 m mesh diameters. Their color-coded frames help differentiate sizes, while its extended tab makes handling easy. They are ready-to-use and each strainer is individually packaged and gamma sterilized.
Catalog No. 50-196-0595 Supplier Cole-Parmer Supplier No. 0439600
Pair these cell strainers directly with your 50-mL centrifuge tubes. Choose from 40, 70 or 100 m mesh diameters. Their color-coded frames help differentiate sizes, while its extended tab makes handling easy. They are ready-to-use and each strainer is individually packaged and gamma sterilized.