Learn More
Chemglass Life Sciences Cell Strainer, 40µm, Individually Wrapped, Color Coded Blue, Sterile
SDP
Catalog No. 501897816
Encompass_Preferred
Cell strainer can be used to obtain uniform single cell suspension which is compatible with 50mL centrifuge tubes of most popular brands on the market. Frames are made from polypropylene with nylon strainers. Individually packaged. Sample filtration. Sample preparation for flow cytometer. Isolate primary cells to consistently obtain a uniform single-cell tissue suspension. Non-pyrogenic, DNase- / RNase-free, non-cytotoxicity.
Catalog No. 50-189-7816 Supplier Chemglass Life Sciences Supplier No. CGN2007040
Cell strainer can be used to obtain uniform single cell suspension which is compatible with 50mL centrifuge tubes of most popular brands on the market. Frames are made from polypropylene with nylon strainers. Individually packaged. Sample filtration. Sample preparation for flow cytometer. Isolate primary cells to consistently obtain a uniform single-cell tissue suspension. Non-pyrogenic, DNase- / RNase-free, non-cytotoxicity.