Learn More
Electron Microscopy Sciences SureStrain™ Cell Strainer w/ Reducing Adapter, 70 µm, White
SDP
Catalog No. 501929303
Encompass_Preferred
The SureStrain Premium Cell Strainer is a fast and easy-to-use system for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh and an unique interlocking stackable design makes tiered filtration possible.
The molded grip and unique geometry ensure aseptic handling. The sterile strainer can be easily removed from the special blister packaging without touching its interior surface or mesh.
The advanced frame/housing design reduces processing time, by eliminating the air-lock effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers. Simply open the sterile package, attach the strainer to the top of a 50 ml tube and add your cell suspension/sample. The provided reducing adapter is for use in 15 ml tubes. Sure Strain produces an optimized filtrate for maximum flow in separate columns and cytometers.
Size: 70 µm
Color: White
Catalog No. 50-192-9303 Supplier Electron Microscopy Sciences Supplier No. 6474902
The SureStrain Premium Cell Strainer is a fast and easy-to-use system for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh and an unique interlocking stackable design makes tiered filtration possible.
The molded grip and unique geometry ensure aseptic handling. The sterile strainer can be easily removed from the special blister packaging without touching its interior surface or mesh.
The advanced frame/housing design reduces processing time, by eliminating the air-lock effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers. Simply open the sterile package, attach the strainer to the top of a 50 ml tube and add your cell suspension/sample. The provided reducing adapter is for use in 15 ml tubes. Sure Strain produces an optimized filtrate for maximum flow in separate columns and cytometers.
Size: 70 µm
Color: White