Learn More
Andwin Scientific CELL STRAINER 70UM 10/CS
SDP
Catalog No. 50727400
Encompass_Preferred
The 70 m Falcon Cell Strainers feature nylon mesh that offers users more even cell suspensions. This product can also be utilized for dissociating clumped cells in separation for cell counting or sub-culture, separating enzymatically dissociated cells from undigested tissue for the initiation of primary cell growing, tissue processing such as fixation, dehydration, and staining.The strainer is suitable to work with 50mL conical tubes. This is sold in a case of 10.
Catalog No. 50-727-400 Supplier Andwin Scientific Supplier No. 6475210
The 70 m Falcon Cell Strainers feature nylon mesh that offers users more even cell suspensions. This product can also be utilized for dissociating clumped cells in separation for cell counting or sub-culture, separating enzymatically dissociated cells from undigested tissue for the initiation of primary cell growing, tissue processing such as fixation, dehydration, and staining.The strainer is suitable to work with 50mL conical tubes. This is sold in a case of 10.