Learn More
Thomas Scientific Cell Strainer, 70um, White, Sterile, Individually Wrapped
SDP
Catalog No. 501461427
Encompass
Cell Strainers Extended tab for easy handling Argos cell strainers offer a simple, ready to use method for securing single-cell suspensions. Available in filter mesh diameters of 40, 70 and 100µm. They are compatible with 50mL centrifuge tubes and feature color-coded polypropylene frames. Individually packaged and gamma sterilized. * Micron dimensions refer to the diameter of the pores.
Catalog No. 50-146-1427 Supplier Thomas Scientific Supplier No. 1170X27
Cell Strainers Extended tab for easy handling Argos cell strainers offer a simple, ready to use method for securing single-cell suspensions. Available in filter mesh diameters of 40, 70 and 100µm. They are compatible with 50mL centrifuge tubes and feature color-coded polypropylene frames. Individually packaged and gamma sterilized. * Micron dimensions refer to the diameter of the pores.