Learn More
CELLTREAT Scientific Products Pestle for Cell Strainer, Individually Wrapped, Sterile
SDP
Catalog No. 50828735
Encompass_Preferred
CELLTREAT Scientific Products Cell Strainer Pestle. The CELLTREAT exclusive pestle design improves manipulation of material in the strainer during use. 5.5 inch pestle length for convenient hand manipulation. Convex pestle head includes molded textured surface to better manipulate slippery material in the cell strainer. RNase/DNase free and non-pyrogenic. Green color. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped. 50/case
Catalog No. 50-828-735 Supplier CELLTREAT Scientific Products Supplier No. 229480
CELLTREAT Scientific Products Cell Strainer Pestle. The CELLTREAT exclusive pestle design improves manipulation of material in the strainer during use. 5.5 inch pestle length for convenient hand manipulation. Convex pestle head includes molded textured surface to better manipulate slippery material in the cell strainer. RNase/DNase free and non-pyrogenic. Green color. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped. 50/case