Learn More
Electron Microscopy Sciences Falcon™ Cell Strainers 35 µm mesh, Sterile
SDP
Catalog No. 501899187
Encompass_Preferred
These nylon meshed devices provide more uniform cell suspensions, and are a perfect tool for separating cells from either clumps or primary tissues. There is no longer a need to fabricate filters from gauze, cotton, paper or fabric. Cell strainers are compatible with 50 ml conical tubes.
Ideal for dissociating clumped cells in separation for cell counting or sub-culture, separating enzymatically dissociated cells from undigested tissue for the initiation of primary cell growing, tissue processing such as fixation, dehydration, and staining. Fits 6 mL tube.
Mesh: 35 µm
Sterile: Sterile
Use: 6 mL tubes
Catalog No. 50-189-9187 Supplier Electron Microscopy Sciences Supplier No. 6475000
These nylon meshed devices provide more uniform cell suspensions, and are a perfect tool for separating cells from either clumps or primary tissues. There is no longer a need to fabricate filters from gauze, cotton, paper or fabric. Cell strainers are compatible with 50 ml conical tubes.
Ideal for dissociating clumped cells in separation for cell counting or sub-culture, separating enzymatically dissociated cells from undigested tissue for the initiation of primary cell growing, tissue processing such as fixation, dehydration, and staining. Fits 6 mL tube.
Mesh: 35 µm
Sterile: Sterile
Use: 6 mL tubes