Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Cell Structure 3D Model Making Kit
SCI_ED

Catalog No. S436046 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S436046 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S436046 Supplier NewPath Learning™ Supplier No. 247739
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Cell Structure 3D Model Making Kit allows students to make four different cell models to learn about subcellular structures.

  • Your students will first make models out of four different cells using the colorful parts provided.
  • Then, they use the models to look at what the subcellular structures do as well as to compare and contrast the different types of cell.
  • Through the process, they will learn about the nucleus, chromosome, mitochondrion, chloroplast, vacuole, cell membrane, cell wall, cytoplasm, ribosomes, plasmids and flagella.
  • The Model Making Kit includes enough materials for students to assemble up to 5 paper models working cooperatively.
  • The comprehensive Teacher Resource Guide included provides illustrated readings, activities, key vocabulary and an assessment, along with teaching opportunities for students to Engage, Explore, Explain, Extend and Evaluate their understanding of the concepts presented.

Specifications

For Use With (Application) Model Assembly
Grade 6 to 10
Includes 5 Models, Teacher Resource Guide, Vinyl Pouch
Length (English) 12 in.
Material Paper
Weight (English) 2 lb.
Width (English) 9 in.
Product Type 3D Model
Grade Level Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.