Learn More
ECM Bioscience Cell Structure Labeling Immunocytochemistry Kit, ECM Biosciences
Catalog No. 502068638
Encompass_Preferred
The cell structure labeling kit can be used to identify important cell structures found in eukaryotic cells. The kit includes a panel of antibodies that detect the following cell structures:Actin Filaments = Phalloidin:FITC Cell Junctions = β-catenin Focal Adhesions= PaxillinIntermediate Filaments = VimentinMicrotubules = α-TubulinNuclear Envelope = Nucleoporin p62 Applications: ICC, IHC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8638 Supplier ECM Bioscience Supplier No. CK7700
The cell structure labeling kit can be used to identify important cell structures found in eukaryotic cells. The kit includes a panel of antibodies that detect the following cell structures:Actin Filaments = Phalloidin:FITC Cell Junctions = β-catenin Focal Adhesions= PaxillinIntermediate Filaments = VimentinMicrotubules = α-TubulinNuclear Envelope = Nucleoporin p62 Applications: ICC, IHC; Species Reactivity: Hu, Rt, Ms