Learn More
Biotium Cell Surface Membrane Dyes Cell and Organelle Stains Cell Surface and Membrane Dyes Chemical Reagents 1/EA
Catalog No. 5000501576
Encompass_Preferred
CytoLiner Fixed Cell Membrane Dyes are novel lipophilic fluorescent dyes developed specifically for use in fixed and mildly permeabilized cells compatible with later antibody staining CytoLiner 495/515 has green fluorescence Kit includes 200 uL of 500X dye and 1 mL of 100X staining buffer
Catalog No. 50-005-01576 Supplier Biotium Supplier No. 30132
CytoLiner Fixed Cell Membrane Dyes are novel lipophilic fluorescent dyes developed specifically for use in fixed and mildly permeabilized cells compatible with later antibody staining CytoLiner 495/515 has green fluorescence Kit includes 200 uL of 500X dye and 1 mL of 100X staining buffer