Learn More
Biotium Cell Surface Membrane Dyes Cell and Organelle Stains Cell Surface and Membrane Dyes Chemical Reagents 1/EA
Supplier:  Biotium 30133
Encompass_Preferred
CytoLiner Fixed Cell Membrane Dyes are novel lipophilic fluorescent dyes developed specifically for use in fixed and mildly permeabilized cells compatible with later antibody staining This red dye has Ex/Em at 570/590 nm Kit includes 200 uL of 500X dye and 1 mL of 100X staining buffer
Catalog No. 50-005-01536