Learn More
BIOLIFE SOLUTIONS LLC Cell Thawing Media 10% Dextran 40 (in 0.9% NaCl) 250mL Bag
SDP
Catalog No. NC0841397
Encompass
This item has a minimum qty of 2 per supplier requirements.
Cell Thawing Media 10% Dextran 40 (in 0.9% NaCl) is manufactured per cGMP guidelines, using USP components and Water for Injection (WFI) quality water. Cell Thawing Media 10% Dextran 40 (in 0.9% NaCl) provides Dextran and saline for washing cryopreserved cells and tissues to dilute or remove cryoprotectants.
Catalog No. NC0841397 Supplier BIOLIFE SOLUTIONS LLC Supplier No. 980203
This item has a minimum qty of 2 per supplier requirements.
Cell Thawing Media 10% Dextran 40 (in 0.9% NaCl) is manufactured per cGMP guidelines, using USP components and Water for Injection (WFI) quality water. Cell Thawing Media 10% Dextran 40 (in 0.9% NaCl) provides Dextran and saline for washing cryopreserved cells and tissues to dilute or remove cryoprotectants.