Learn More
Epoch Life Science Inc GenCatch™ Total RNA Extraction System (50 preps) Uniquely formulated buffers, reinforced columns with collection tubes, along with protocols.
SDP
Catalog No. 501538096
Encompass_Preferred
Provides a simple, fast and cost-effective method to purify total RNA from various starting materials (such as cultured cells, tissues, bacteria, yeast, etc.). Total RNA with a molecular size greater than 200 nucleotides (which excludes smaller RNA such as 5S and 5.8S RNA, and tRNA) can be isolated. Silica membrane technology. Up to 65 ug yield. 200 ul elution volume. No phenol/chloroform extraction nor ethanol precipitation required. Instructions for sample type listed in protocol. Used by labs around the world. 18 month shelf life.
Catalog No. 50-153-8096 Supplier Epoch Life Science Inc Supplier No. 1660050
Provides a simple, fast and cost-effective method to purify total RNA from various starting materials (such as cultured cells, tissues, bacteria, yeast, etc.). Total RNA with a molecular size greater than 200 nucleotides (which excludes smaller RNA such as 5S and 5.8S RNA, and tRNA) can be isolated. Silica membrane technology. Up to 65 ug yield. 200 ul elution volume. No phenol/chloroform extraction nor ethanol precipitation required. Instructions for sample type listed in protocol. Used by labs around the world. 18 month shelf life.