Learn More
CELLTREAT Scientific Products 656cm2 Tissue Culture Treated Cell Tower, 1 Layer, Vent Cap, Sterile
SDP
Catalog No. NC3377716
Encompass_Preferred
Tissue Culture Treated Cell Tower system scales up standard Tissue Culture Flasks to maximize adherent cell yieldsManufactured from pure, optically clear polystyrene, providing a clear visualization of cell culturesEfficient design facilitates up to 6416cm2 culture area while reducing plastic material usage compared to traditional flasksLarge, internal channels allow free media flow to reduce foamingAvoid contamination risks and save time and resources by culturing higher yields in fewer vesselsIdeal for growing and harvesting cellular products, such as antibodies and therapeutic compounds
Catalog No. NC3377716 Supplier CELLTREAT Scientific Products Supplier No. 229301
Tissue Culture Treated Cell Tower system scales up standard Tissue Culture Flasks to maximize adherent cell yieldsManufactured from pure, optically clear polystyrene, providing a clear visualization of cell culturesEfficient design facilitates up to 6416cm2 culture area while reducing plastic material usage compared to traditional flasksLarge, internal channels allow free media flow to reduce foamingAvoid contamination risks and save time and resources by culturing higher yields in fewer vesselsIdeal for growing and harvesting cellular products, such as antibodies and therapeutic compounds