Learn More
Mitegen Stainless Steel 100 Cell Capacity 5 Shelf Inventory Rack For IC Biomedical CXR500 Dryshipper
SDP
Catalog No. NC3366167
Encompass
Stainless Steel 5-Shelf Inventory Rack, Holds 100-Cell Boxes (5.25"x5.25"x2" Max), For CXR500 Dryshipper. Cell boxes sold separately.
Catalog No. NC3366167 Supplier Mitegen Supplier No. TWRS309C44
Stainless Steel 5-Shelf Inventory Rack, Holds 100-Cell Boxes (5.25"x5.25"x2" Max), For CXR500 Dryshipper. Cell boxes sold separately.