Learn More
Partec North America Sterile single-pack CellTrics™ filters (50/box) (Yellow)
Catalog No. NC9491906
Encompass
CellTrics can be used to filter any kind of liquid suspension, e.g. for isolation of cells and nuclei from cell debris and aggregates and also for filtering either microorganisms or cell cultures
Catalog No. NC9491906 Supplier Partec North America Supplier No. 040042327
CellTrics can be used to filter any kind of liquid suspension, e.g. for isolation of cells and nuclei from cell debris and aggregates and also for filtering either microorganisms or cell cultures