Learn More
Cell Viability Incorporated CELL VIABILITY REAGENTS
Catalog No. 501351687
Encompass
Cell Viability reagent kit designed for use on automated cell viability analyzers. Four reagent pack with 4 x 200 sample vials.
Catalog No. 501351687 Supplier Cell Viability Incorporated Supplier No. 2400101
Cell Viability reagent kit designed for use on automated cell viability analyzers. Four reagent pack with 4 x 200 sample vials.