Learn More
Biolegend Cell-Vive™ GMP Ultra-LEAF™ Biotin anti-human CD34 SF [581]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg
Supplier:  Biolegend 343549
Encompass_Preferred
Cell-Vive™ GMP Ultra-LEAF™ Biotin anti-human CD34 SF [581]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg
Catalog No. 50-272-3290