Learn More
Med Vet International Cell-VU Hemacytometer System Kit, J0821X
Catalog No. 5000582146
Encompass_Preferred
The safety hemacytometer system for use with all manual-counting procedures.Sperm • CSFWBC • UrineConsists of both the slide and cover slipsSlide is dual chambered providing for two determinations per sampleCover slips are laser-etched grid that is easy to view
Catalog No. 50-005-82146 Supplier Med Vet International Supplier No. JOR821X
The safety hemacytometer system for use with all manual-counting procedures.Sperm • CSFWBC • UrineConsists of both the slide and cover slipsSlide is dual chambered providing for two determinations per sampleCover slips are laser-etched grid that is easy to view