Learn More
Starna Cells Inc CELL W/CAP STAND
SDP
Catalog No. NC9960553
Encompass_Preferred
Cell, Rectangular; Standard; Starna; w/septum cap; Window and Wall Thickness: 1.25mm; Polished windows: 2; Pathlength: 10mm; Window material: Spectrosil Quartz; Useable Range: 170-2700nm; Nominal Vol.: 3.5 mL
Catalog No. NC9960553 Supplier Starna Cells Inc Supplier No. 1Q10GL14S
Cell, Rectangular; Standard; Starna; w/septum cap; Window and Wall Thickness: 1.25mm; Polished windows: 2; Pathlength: 10mm; Window material: Spectrosil Quartz; Useable Range: 170-2700nm; Nominal Vol.: 3.5 mL