Learn More
Aqua Solutions Cell Wash Standard #1 20.0 ppm Ca, Fe, Mg in 10% v/v HNO₃ (500ML)
SDP
Catalog No. NC9885869
Encompass_Preferred
Cell Wash Standard #1 20.0 ppm Ca, Fe, Mg in 10% v/v HNO₃ (500ML)
Catalog No. NC9885869 Supplier Aqua Solutions Supplier No. DCP873135
Cell Wash Standard #1 20.0 ppm Ca, Fe, Mg in 10% v/v HNO₃ (500ML)