Learn More
STEMCELL Technologies CellAdhere™ Collagen I-Coated, 24-Well Flat-Bottom Plate, Size: 5 Units
SDP
Catalog No. 502045526
Encompass
CellAdhereᵀᴹ Collagen I-Coated, Flat-Bottom Plates are sterile, clear polystyrene plates. For optimal cell attachment, these plates have been coated with high-purity collagen I derived from rat tail tendon. These plates can be used with a variety of cell types and cell culture applications.
Catalog No. 50-204-5526 Supplier STEMCELL Technologies Supplier No. 1000364
CellAdhereᵀᴹ Collagen I-Coated, Flat-Bottom Plates are sterile, clear polystyrene plates. For optimal cell attachment, these plates have been coated with high-purity collagen I derived from rat tail tendon. These plates can be used with a variety of cell types and cell culture applications.