Learn More
STEMCELL Technologies CellAdhere™ Dilution Buffer, Size: 100 mL
SDP
Catalog No. NC0672885
Encompass
Use CellAdhere™ Dilution Buffer in combination with Vitronectin XF™ (https://www.stemcell.com/vitronectin-xf.html) (developed and manufactured by Nucleus Biologics) to provide a defined, xeno-free surface for the culture of human embryonic stem (ES) and induced pluripotent stem (iPS) cells.CellAdhere™ is required for use with Vitronectin XF™, which may be used as an alternative to Corning Matrigel to support the growth and differentiation of human pluripotent stem cells.
Catalog No. NC0672885 Supplier STEMCELL Technologies Supplier No. 07183
Use CellAdhere™ Dilution Buffer in combination with Vitronectin XF™ (https://www.stemcell.com/vitronectin-xf.html) (developed and manufactured by Nucleus Biologics) to provide a defined, xeno-free surface for the culture of human embryonic stem (ES) and induced pluripotent stem (iPS) cells.CellAdhere™ is required for use with Vitronectin XF™, which may be used as an alternative to Corning Matrigel to support the growth and differentiation of human pluripotent stem cells.