Learn More
Takara Bio ellartis DEF-CS™ 500 Culture System, 1 kit
SDP
Catalog No. NC1114880
Encompass
Cellartis DEF-CS 500 Culture System is a defined culture system for efficient expansion of undifferentiated human pluripotent stem cells. This kit includes basal medium, coating substrate, and additives.
Catalog No. NC1114880 Supplier Takara Bio Supplier No. Y30010
Cellartis DEF-CS 500 Culture System is a defined culture system for efficient expansion of undifferentiated human pluripotent stem cells. This kit includes basal medium, coating substrate, and additives.