Learn More
InVitria Cellastim S - Recombinant Human Serum Albumin - 100g
Catalog No. 501354696
Encompass
InVitria’s Cellastim S recombinant human serum albumin (rHSA) is completely animal- and blood-free and is optimized to enhance the performance in cell culture media due to the favorable lipid profile. Cellastim S is compatible with a variety of different cell types including immune cells, VERO, MSC, HEK, CHO, etc.InVitria manufactures three animal- and blood-free rHSA components: Cellastim S, Exbumin™, and Optibumin. Cellastim S is optimized for cell culture applications. Exbumin is a regulatory approved excipient in final formulations. Optibumin is the highest-purity rHSA available on the market. Made in the USA.
Catalog No. 50-135-4696 Supplier InVitria Supplier No. 777HSA017S100G
InVitria’s Cellastim S recombinant human serum albumin (rHSA) is completely animal- and blood-free and is optimized to enhance the performance in cell culture media due to the favorable lipid profile. Cellastim S is compatible with a variety of different cell types including immune cells, VERO, MSC, HEK, CHO, etc.InVitria manufactures three animal- and blood-free rHSA components: Cellastim S, Exbumin™, and Optibumin. Cellastim S is optimized for cell culture applications. Exbumin is a regulatory approved excipient in final formulations. Optibumin is the highest-purity rHSA available on the market. Made in the USA.