Description
Faster slide reviews
- CellaVision Digital Morphology Solutions allow you to reduce turnaround times with automated pre-classification of the WBC differential, RBC morphology and platelet estimates.
- Remote review expedites expert opinions, making more efficient use of experienced morphologists; time.
Standardized differential results
- Automatic cell location and pre-classification reduce the subjectivity of cell identification and improve the quality of results.
Remote slide reviews
- Cell images are stored in a centralized database where they can be accessed, reviewed and reported on from any laboratory or physician's office location within the network.
Integrated with LIS
- Cell images can be stored with patient records, making it easy to track patient progress over time.
Enhance ergonomics
- CellaVision Digital Morphology solutions analyzers do not cause neck, wrist or eye fatigue like microscopy does. As a result, you can help preserve your staff;s health and improve their satisfaction.
Specifications
Specifications
| For Use With (Equipment) | Beckman Coulter Analyzers |
| Throughput | 20 Slides/hr. |