Learn More
Amsbio LLC CELLBANKER 1 Cryopreservation Media (100ml)(formerly 11888)
Catalog No. 502566888
Encompass
The CELLBANKER series of cell freezing media allows for the stable long-term storage of cells. With its unique formulation which enables stable cryopreservation and high viability after freeze-thaw procedures, CELLBANKER is a trusted solution for the storage of any cell type including sensitive cell lines, and is cited in over 1600 publications. CELLBANKER 1 MAIN FEATURES: Ready-to-use serum-containing cell cryopreservation medium | Suitable for a range of mammalian cell cultures, organoids, and tissue | Simple protocol with consistent high cell viability post-thaw (>90% for many cell lines) | Direct freezing at -80°C with no need for programmable freezer or liquid nitrogen | Tested on a wide range of cells and highly cited | No programmed freezer or liquid nitrogen required | Long shelf life (3 years from manufacturing date). Contains serum, DMSO, glucose, salts and buffer.
Catalog No. 50-256-6888 Supplier Amsbio LLC Supplier No. 11910
The CELLBANKER series of cell freezing media allows for the stable long-term storage of cells. With its unique formulation which enables stable cryopreservation and high viability after freeze-thaw procedures, CELLBANKER is a trusted solution for the storage of any cell type including sensitive cell lines, and is cited in over 1600 publications. CELLBANKER 1 MAIN FEATURES: Ready-to-use serum-containing cell cryopreservation medium | Suitable for a range of mammalian cell cultures, organoids, and tissue | Simple protocol with consistent high cell viability post-thaw (>90% for many cell lines) | Direct freezing at -80°C with no need for programmable freezer or liquid nitrogen | Tested on a wide range of cells and highly cited | No programmed freezer or liquid nitrogen required | Long shelf life (3 years from manufacturing date). Contains serum, DMSO, glucose, salts and buffer.