Learn More
Iwai North America CELLBANKER 1: Ready to use cryopreservation media enriched with newborn calf serum. Size: 20mL
Catalog No. NC2732983
Encompass_Preferred
CELLBANKER 1 is a flagship in the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, it contains newborn calf serum, is ready to use, and requires no liquid nitrogen for cell preservation. With over a decade of data and extensive testing on diverse cell lines, tissues, and organoids, trust CELLBANKER 1 for safe, hassle-free cell storage.
Catalog No. NC2732983 Supplier Iwai North America Supplier No. 11911
CELLBANKER 1 is a flagship in the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, it contains newborn calf serum, is ready to use, and requires no liquid nitrogen for cell preservation. With over a decade of data and extensive testing on diverse cell lines, tissues, and organoids, trust CELLBANKER 1 for safe, hassle-free cell storage.