Learn More
Iwai North America CELLBANKER 2: Ready to use serum free cryopreservation media. Size: 100mL
Catalog No. NC2732993
Encompass
CELLBANKER 2 is a part of the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, it is serum free, is ready to use, and requires no liquid nitrogen for cell preservation. With over a decade of data and extensive testing on diverse cell lines, tissues, and organoids, trust CELLBANKER 2 for safe, hassle-free cell storage.
Catalog No. NC2732993 Supplier Iwai North America Supplier No. 11914
CELLBANKER 2 is a part of the renowned CELLBANKER series, cited in over 1400 publications. As part of Japan's most trusted line of cryopreservation media, it is serum free, is ready to use, and requires no liquid nitrogen for cell preservation. With over a decade of data and extensive testing on diverse cell lines, tissues, and organoids, trust CELLBANKER 2 for safe, hassle-free cell storage.