Learn More
Corning™ CellBIND™ 384-Well, CellBIND-Treated, Flat-Bottom, Low Flange Microplate
GSA_VAGREENER_CHOICE
Catalog No. 07201015 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Catalog No. Color
07-201-015 Black, Clear Bottom
1 options
This item is not returnable. View return policy
Catalog No. 07-201-015 Supplier Corning™ Supplier No. 3770
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Culture Area 0.06 cm2
Array 16 x 24
Packaging 20/Pk., 100/Cs.
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Sterile
Surface Treatment CellBIND
Well Shape Square, Flat Bottom (F-well)
No. per Case 100/Cs.
No. per Pack 20/Pk.
Quantity 100/Cs.