Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CellBIND™ HYPERStack™ Cell Culture Vessels - New Design
GREENER_CHOICE

Catalog No. 10320186 Shop All Corning Products
Change view
Click to view available options
Culture Area:
18,000 cm2
6000 cm2
Product Type:
12-layer vessel
36-layer vessel
No. per Case:
2/Cs.
4/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Product Type No. per Case
10-320-186 18,000 cm2 36-layer vessel 2/Cs.
10-320-184 6000 cm2 12-layer vessel 4/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-320-186 Supplier Corning™ Supplier No. 20036
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint.

Corning's High Yield PERformance (HYPER) Platform - Closed System for High Yield Cell Growth

The Corning HYPERStack cell culture vessel combines the best of two Corning products: the Corning CellSTACK™ and the Corning HYPERFlask™ cell culture vessels. The utilization of gas-permeable film technology provided in the spatial footprint of the CellSTACK vessel allows the HYPERStack platform to be among the most efficient, scalable cell culture vessels for adherent cell culture available today.

Features and Benefits

  • More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint
  • Closed system – no open fluid manipulations
  • Scalable product – multiple size offerings support scale-up and scale-out
  • Ergonomic design – easier manipulation with handling equipment and accessories
  • Fixed media volume – 0.2 mL/cm2 fills vessel for less volumetric waste

Surface Treatment Guide

Corning CellBIND™ surface treatment is designed to enhance cell attachment and increase cell yields, improving attachment of loosely adherent cells and maintaining growth in reduced-serum or serum-free conditions. Alternatively, a non-treated option can support conventional cell attachment and allow for a faster and gentler harvest procedure for downstream processing and assays.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN ISO 13485, FDA CFR 21 Part 820, cGMP, BSE/TSE (EMA/410/01), Non-Pyrogenic, USP Class VI Tested, Radiation Sterilized (ANSI/AAMI/ISO 11137), SAL 10⁻⁶
Capacity (Metric) 3920 mL
Material Polystyrene
Height (English) 10.97 in.
Height (Metric) 278 mm
Length (English) 13.42 in.
Length (Metric) 342 mm
No. per Case 2/Cs.
No. per Pack 1 Ea.
Product Type 36-layer vessel
Shape Rectangular
Width (English) 8.15 in.
Width (Metric) 207 mm
Sterility Sterile
Bottom Shape Rectangular
Culture Area 18,000 cm2
Surface Treatment CellBIND
Volume (Metric) Working 3920 mL
Tray Layers 36
Quantity 1 Ea.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.