Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellBlox™ Blocking Buffer

Catalog No. B001T06F01
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
500 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B001T06F01 500 Tests
B001T02F01 25 Tests
B001T03F01 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B001T06F01 Supplier Invitrogen™ Supplier No. B001T06F01
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Formulated to block nonspecific binding of Invitrogen™ NovaFluor™ labels with cells, which can result in higher background labeling. Also recommended for cyanine-based dyes or cyanine-based tandem dyes to block non-specific interactions with monocytes and macrophages to minimize background labeling.

  • Formulated to block nonspecific binding of Invitrogen™ NovaFluor™ labels with cells that can result in higher background labeling
  • CellBlox™ Blocking Buffer is a non-antibody, non-protein–based blocking solution, and should be used every time a NovaFluor dye is used for labeling any cell type to minimize background labeling
  • Recommended for use with cyanine-based dyes or cyanine-based tandem dyes to block non-specific interactions with monocytes and macrophages to minimize background labeling
  • Use of CellBlox Blocking Buffer requires minimal change to most flow cytometry staining protocols
  • Add 5 μL of buffer directly to a cell suspension containing 103 to 108 cells prior to the addition of an antibody, with 100 μL as a final staining volume
  • Buffer may be added to an antibody cocktail mixture prior to labeling cells, by adding 5 μL of buffer for every stained sample to be labeled with the antibody cocktail mixture, with 100 μL as a final staining volume
  • Always use CellBlox Blocking Buffer with NovaFluor dyes when labeling cells for best background reduction
  • Compatible with all fluorophores and with Invitrogen™ LIVE/DEAD™ Fixable Dead Cell Stains
  • Can be used with any fluorophore-antibody conjugate as a high-performance monocyte and macrophage blocking solution
  • CellBlox Blocking Buffer is compatible with other blocking reagents such as Fc Block, blocking proteins, Brilliant Stain Buffer, and Super Bright Complete Staining Buffer
  • Buffer is not required when labeling antibody-capture beads
TRUSTED_SUSTAINABILITY

Specifications

Description Blocking Buffer
For Use With (Application) Flow Cytometry
Quantity 500 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.