Learn More
Biotium CellBrite Fix 488, 5 vials
Catalog No. NC1700370
Encompass_Preferred
Unique fluorogenic membrane dyes that covalently stain the plasma membrane in live cells, and can withstand both fixation and detergent permeabilization.
Catalog No. NC1700370 Supplier Biotium Supplier No. 30090
Unique fluorogenic membrane dyes that covalently stain the plasma membrane in live cells, and can withstand both fixation and detergent permeabilization.