Learn More
Biotium CellBrite™ Fix 640 Membrane Labeling Kit
Supplier:  Biotium 30089
Encompass_Preferred
CellBrite Fix 640 is a far-red fluorescent reactive dye that covalently labels the surface of live cells. Unlike other membrane dyes, surface staining is well-retained after permeabilization or methanol fixation, for subsequent immunostaining. Ex/Em 638/667 nm. Each vial makes 20 uL of 1000X dye.
Catalog No. NC1622830