Learn More
Biotium CELLBRITE GREEN CYTOPLASMIC
Supplier:  Biotium 30021
Encompass_Preferred
Biotium’s CellBrite™ Cytoplasmic Membrane Labeling Dyes are lipophilic carbocyanine dyes that label cytoplasmic membrane and intracellular membrane structures efficiently and permanently. They have been used as tracers in cell–cell fusion, cellular adhesion, and migration applications due to their properties of low cytotoxicity and high resistance to intercellular transfer. CellBrite™ dyes are dye delivery solutions that can be added directly to normal culture media to label suspended or adherent cells in culture. CellBrite™ Green Cytoplasmic Membrane Labeing Dye features green fluorescent Neuro-DiO, which has excitaiton/emission at 484/501 nm in membranes. Biotium offers a selection of CellBrite™ dyes to allow cell populations to be marked in distinctive fluorescent colors for identification after mixing. Double labeling can identify cells that have fused or formed stable clusters. 1 mL labeling solution.
Catalog No. NC1136383