Learn More
Biotium CellBrite Cytoplasmic Membrane Dyes
Catalog No. NC1342358
Encompass_Preferred
CellBrite Cytoplasmic Membrane Dyes are a set of fluorescent membrane dyes used to stain the plasma membrane of live or fixed cells for microscopy applications. They produce bright staining of the plasma membrane with low background fluorescence. The dyes are available in green, orange, deep red, and near-infrared fluorescent colors. They are compatible with common filter sets and have high photostability. The dyes can be used for various applications including cell tracking, cell proliferation studies, and co-localization experiments. They are supplied as DMSO solutions and are non-toxic to cells at working concentrations.
Catalog No. NC1342358 Supplier Biotium Supplier No. 30023
CellBrite Cytoplasmic Membrane Dyes are a set of fluorescent membrane dyes used to stain the plasma membrane of live or fixed cells for microscopy applications. They produce bright staining of the plasma membrane with low background fluorescence. The dyes are available in green, orange, deep red, and near-infrared fluorescent colors. They are compatible with common filter sets and have high photostability. The dyes can be used for various applications including cell tracking, cell proliferation studies, and co-localization experiments. They are supplied as DMSO solutions and are non-toxic to cells at working concentrations.