Learn More
Revvity Health Sciences Inc PhenoPlate 96-well, black, optically clear flat-bottom, ultra-low-attachment-coated, lids, case of 10
Catalog No. 502105653
Encompass
PhenoPlate 96-well, black, optically clear flat-bottom, ultra-low-attachment-coated, lids, case of 10
Catalog No. 50-210-5653 Supplier Revvity Health Sciences Inc Supplier No. 6055800
PhenoPlate 96-well, black, optically clear flat-bottom, ultra-low-attachment-coated, lids, case of 10