Learn More
Revvity Health Sciences Inc PhenoPlate 384-well, black, optically clear flat-bottom, tissue-culture treated, lids, case of 50
Catalog No. 502099109
Encompass
PhenoPlate 384-well, black, optically clear flat-bottom, tissue-culture treated, lids, case of 50
Catalog No. 50-209-9109 Supplier Revvity Health Sciences Inc Supplier No. 6057300
Add to Cart
Add to Cart
PhenoPlate 384-well, black, optically clear flat-bottom, tissue-culture treated, lids, case of 50