Learn More
Revvity Health Sciences Inc PhenoPlate 384-well, black, optically clear flat-bottom, tissue-culture treated, non-irradiated, lids, case of 160
Supplier:  Revvity Health Sciences Inc 6057328
Encompass
PhenoPlate 384-well, black, optically clear flat-bottom, tissue-culture treated, non-irradiated, lids, case of 160
Catalog No. 50-211-9443