Learn More
Greiner Bio-One CELLCOAT™ 384-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000046 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Catalog No. Color
07-000-046 Black, Clear Bottom
1 options
Catalog No. 07-000-046 Supplier Greiner Bio-One Supplier No. 781946
Add to Cart
Edge
Add to Cart
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
  • Improves adhesion of different cell types to polystyrene surfaces
  • Reduced-serum or serum-free cultivation
  • Increase in isolation and cultivation efficiency
  • Ready-to-use products, time-saving
  • Consistent quality
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Array 16 x 24
Autoclavable Not Autoclavable
Packaging 5/Pk.
Color Black, Clear Bottom
Product Type Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Non-sterile
Surface Treatment Poly-D-Lysine
Well Shape Rounded Square
Max. RCF 4800 x g (swinging-bucket rotor)
No. per Case 20
No. per Pack 5
Quantity 20/Cs.
Temperature Range (Metric) Room Temperature