Learn More
Greiner Bio-One CELLCOAT™ 384-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Shop All Greiner Bio-One Products

Description
- Improves adhesion of different cell types to polystyrene surfaces
- Reduced-serum or serum-free cultivation
- Increase in isolation and cultivation efficiency
- Ready-to-use products, time-saving
- Consistent quality
Specifications
Specifications
| Barcode | No Barcode |
| Lid | With Lid |
| Array | 16 x 24 |
| Autoclavable | Not Autoclavable |
| Color | Black |
| Product Type | Cell Culture Microplate |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 15 to 110 μL |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.