Learn More
Greiner Bio-One CELLCOAT™ 384-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000897 Shop All Greiner Bio-One Products
Change view
Click to view available options
Packaging:
20/Pk., 120/Cs.
5/Pk., 20/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
07-000-897 20/Pk., 120/Cs.
07-000-895 5/Pk., 20/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-897 Supplier Greiner Bio-One Supplier No. 781948
Add to Cart
Add to Cart
Request a Quote
Color: Clear; Sterility: Non-sterile; Lid: With Lid
  • Improves adhesion of different cell types to polystyrene surfaces
  • Reduced-serum or serum-free cultivation
  • Increase in isolation and cultivation efficiency
  • Ready-to-use products, time-saving
  • Consistent quality
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Array 16 x 24
Autoclavable Not Autoclavable
Packaging 20/Pk., 120/Cs.
Color Clear
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Non-sterile
Surface Treatment Poly-D-Lysine
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 120
No. per Pack 20
Temperature Range (Metric) Room Temperature